1-(3-Hydroxyphenyl)piperazine 97%

Leverandør: Apollo Scientific
Warning

Synonymer: 3-(Piperazin-1-yl)phenol, m-(1-Piperazinyl)phenol, 3-(1-Piperazinyl)phenol

OR6853-250MG
APOSOR6853-250MGEA 270 NOK
APOSOR6853-250MG
1-(3-Hydroxyphenyl)piperazine 97%
1-(3-Hydroxyphenyl)piperazine
Formel: C₁₀H₁₄N₂O
MW: 178,23 g/mol
Smeltepunkt: 210 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00052896
CAS-nummer: 59817-32-2

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR