4-Chloro-6-nitro-o-toluidine 98%

Leverandør: Apollo Scientific
Warning

OR6914-250MG OR6914-5G
APOSOR6914-250MGEA 270 NOK
APOSOR6914-250MG APOSOR6914-5G
4-Chloro-6-nitro-o-toluidine 98%
4-Chloro-6-nitro-o-toluidine
Formel: C₇H₇ClN₂O₂
Smeltepunkt: 121…123 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD01320687
CAS-nummer: 62790-50-5

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR