3,4'-Dimethylbiphenyl

Leverandør: Apollo Scientific
Warning

OR70103-1G OR70103-5G
APOSOR70103-1GEA 680 NOK
APOSOR70103-1G APOSOR70103-5G
3,4'-Dimethylbiphenyl
3,4'-Dimethylbiphenyl
Formel: C₁₄H₁₄
MW: 182.261 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00602700
CAS-nummer: 7383-90-6

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR