2,3'-Dimethylbiphenyl

Leverandør: Apollo Scientific
Warning

OR70105-1G OR70105-250MG
APOSOR70105-1GEA 3150 NOK
APOSOR70105-1G APOSOR70105-250MG
2,3'-Dimethylbiphenyl
2,3'-Dimethylbiphenyl
Formel: C₁₄H₁₄
MW: 182.261 g/mol
Kokepunkt: 273…274 °C
Tetthet: 0.9984 g/cm³ (25 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD22628915
CAS-nummer: 611-43-8

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR