3,3'-Dimethylbiphenyl

Leverandør: Apollo Scientific
Warning

Synonymer: 3,3'-Bitolyl, 3,3'-Dimethyl-1,1'-biphenyl

OR70107-1G OR70107-5G
APOSOR70107-1GEA 485 NOK
APOSOR70107-1G APOSOR70107-5G
3,3'-Dimethylbiphenyl
3,3'-Dimethylbiphenyl
Formel: C₁₄H₁₄
MW: 182.27 g/mol
Kokepunkt: 286 °C/713mmHg
Smeltepunkt: 44382 °C
Tetthet: 0.999 g/cm³ (25 °C)
Flash punkt: 113 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008534
CAS-nummer: 612-75-9

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR