4-[1,2,4]Triazol-1-ylbutyric acid

Leverandør: Apollo Scientific
Warning

OR7105-1G
APOSOR7105-1GEA 1975 NOK
APOSOR7105-1G
4-[1,2,4]Triazol-1-ylbutyric acid
4-[1,2,4]Triazol-1-ylbutyric acid
Formel: C₆H₉N₃O₂
MW: 155,16 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD06801317
CAS-nummer: 158147-52-5

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR