3-Bromo-p-toluidine 99%

Leverandør: Apollo Scientific
Danger

Synonymer: 4-Amino-2-bromotoluene, 3-Bromo-4-methylaniline

OR7118-100G OR7118-250G OR7118-500G
APOSOR7118-100GEA 790 NOK
APOSOR7118-100G APOSOR7118-250G APOSOR7118-500G
3-Bromo-p-toluidine 99%
3-Bromo-p-toluidine
Formel: C₇H₈BrN
Kokepunkt: 254…257 °C
Smeltepunkt: 25…30 °C
Flash punkt: 113 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00134176
CAS-nummer: 7745-91-7
EINECS: 231-807-8
FN: 2811
ADR: 6.1,III

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR