1,7,7-Trimethylbicyclo[2.2.1]heptane 95+%

Leverandør: Apollo Scientific
Warning

Synonymer: Bornane, Camphane

OR7980-250MG
APOSOR7980-250MGEA 0 NOK
APOSOR7980-250MG
1,7,7-Trimethylbicyclo[2.2.1]heptane 95+%
1,7,7-Trimethylbicyclo[2.2.1]heptane
Formel: C₁₀H₁₈
MW: 138,25 g/mol
Smeltepunkt: 156…159 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00213517
CAS-nummer: 464-15-3

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR