2-Chloro-4-cyanobenzenesulphonyl chloride 95+%

Leverandør: Apollo Scientific
Danger

Synonymer: 2-Chloro-4-cyanobenzene-1-sulphonyl chloride

OR8053-1G
APOSOR8053-1GEA 270 NOK
APOSOR8053-1G
2-Chloro-4-cyanobenzenesulphonyl chloride 95+%
2-Chloro-4-cyanobenzenesulphonyl chloride
Formel: C₇H₃Cl₂NO₂S
MW: 236,08 g/mol
Smeltepunkt: 101…106 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00177089
CAS-nummer: 254749-11-6
FN: 3261
ADR: 8,III

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR