2-[(7-Chloroquinolin-4-yl)thio]acetohydrazide 97%

Leverandør: Apollo Scientific
Warning

OR8064-1G
APOSOR8064-1GEA 840 NOK
APOSOR8064-1G
2-[(7-Chloroquinolin-4-yl)thio]acetohydrazide 97%
2-[(7-Chloroquinolin-4-yl)thio]acetohydrazide
Formel: C₁₁H₁₀ClN₃OS
Smeltepunkt: 213…216 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD02180374
CAS-nummer: 306935-50-2

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR