3',2,2-Trimethylpropiophenone 95%

Leverandør: Apollo Scientific

Synonymer: 2,2-Dimethyl-1-m-tolylpropan-1-one

OR900478-1G
APOSOR900478-1GEA 2600 NOK
APOSOR900478-1G
3',2,2-Trimethylpropiophenone 95%
3',2,2-Trimethylpropiophenone
Formel: C₁₂H₁₆O
MW: 176.26 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD03841163
CAS-nummer: 50390-49-3

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR