1H-Pyrazolo[3,4-b]pyrazin-3-amine

Leverandør: Apollo Scientific
Warning

OR902364-1G
APOSOR902364-1GEA 770 NOK
APOSOR902364-1G
1H-Pyrazolo[3,4-b]pyrazin-3-amine
1H-Pyrazolo[3,4-b]pyrazin-3-amine
Formel: C₅H₅N₅
MW: 135.1300 g/mol
Smeltepunkt: 244…246 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD09909663
CAS-nummer: 81411-64-5

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR