Nicotinamidine hydrochloride

Leverandør: Apollo Scientific
Warning

OR9026-1G OR9026-10G
APOSOR9026-1GEA 270 NOK
APOSOR9026-1G APOSOR9026-10G
Nicotinamidine hydrochloride
Nicotinamidine hydrochloride
Formel: C₆H₈ClN₃
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00054326
CAS-nummer: 7356-60-7

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR