2-Cyano-4-methylpyridine 97%

Leverandør: Apollo Scientific
Danger

Synonymer: 4-Methylpyridine-2-carbonitrile, 4-Methylpicolinonitrile, 2-Cyano-γ-picoline, 2-Cyano-4-picoline, 4-Methyl-2-pyridinecarbonitrile

OR9027-5G
APOSOR9027-5GEA 270 NOK
APOSOR9027-5G
2-Cyano-4-methylpyridine 97%
2-Cyano-4-methylpyridine
Formel: C₇H₆N₂
MW: 118,14 g/mol
Kokepunkt: 145…148 °C/38mmHg
Smeltepunkt: 83…87 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00128868
CAS-nummer: 1620-76-4
FN: 3439
ADR: 6.1,III

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR