1-Methyl-2,2'-bipiperidine 97%

Leverandør: Apollo Scientific

OR904408-1G
APOSOR904408-1GEA 7300 NOK
APOSOR904408-1G
1-Methyl-2,2'-bipiperidine 97%
1-Methyl-2,2'-bipiperidine
Formel: C₁₁H₂₂N₂
MW: 182.306 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD02663572
CAS-nummer: 118046-22-3

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR