(2-Ethoxy-4-formylphenyl)boronic acid 96%

Leverandør: Apollo Scientific

OR905950-1G
APOSOR905950-1GEA 1500 NOK
APOSOR905950-1G
(2-Ethoxy-4-formylphenyl)boronic acid 96%
(2-Ethoxy-4-formylphenyl)boronic acid
Formel: C₉H₁₁BO₄
MW: 193.992 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD22383668
CAS-nummer: 2253871-63-3

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR