α-Tropolone 98%

Leverandør: Apollo Scientific

OR906152-100G
APOSOR906152-100GEA 2000 NOK
APOSOR906152-100G
α-Tropolone 98%
α-Tropolone
Formel: C₇H₆O₂
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004158
CAS-nummer: 533-75-5

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR