Methyl quinoline-3-carboxylate 95%

Leverandør: Apollo Scientific

OR906465-1G OR906465-5G
APOSOR906465-1GEA 1475 NOK
APOSOR906465-1G APOSOR906465-5G
Methyl quinoline-3-carboxylate 95%
Methyl quinoline-3-carboxylate
Formel: C₁₁H₉NO₂
MW: 187.2 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD03550859
CAS-nummer: 53951-84-1

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR