1-Ethyl-1H-pyrazol-5-ylboronic acid 95%

Leverandør: Apollo Scientific

Synonymer: (1-Ethyl-1H-pyrazol-5-yl)boronic acid

OR907353-5G
APOSOR907353-5GEA 2375 NOK
APOSOR907353-5G
1-Ethyl-1H-pyrazol-5-ylboronic acid 95%
1-Ethyl-1H-pyrazol-5-ylboronic acid
Formel: C₅H₉BN₂O₂
MW: 139.948 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD03419491
CAS-nummer: 1095080-54-8

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR