Methyl 2-methylbenzimidazole-5-carboxylate 97%

Leverandør: Apollo Scientific

OR909478-1G
APOSOR909478-1GEA 5375 NOK
APOSOR909478-1G
Methyl 2-methylbenzimidazole-5-carboxylate 97%
Methyl 2-methylbenzimidazole-5-carboxylate
Formel: C₁₀H₁₀N₂O₂
MW: 190,199 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD19685000
CAS-nummer: 106429-51-0

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR