N-Methylpiperidin-3-amine dihydrochloride 97%

Leverandør: Apollo Scientific

Synonymer: 3-(Methylamino)piperidine dihydrochloride

OR910998-100G
APOSOR910998-100GEA 1450 NOK
APOSOR910998-100G
N-Methylpiperidin-3-amine dihydrochloride 97%
N-Methylpiperidin-3-amine dihydrochloride
Formel: C₆H₁₆Cl₂N₂
MW: 187.1100 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00673143
CAS-nummer: 127294-77-3

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR