(3-Propoxyphenyl)acetic acid 98%

Leverandør: Apollo Scientific

OR911179-1G OR911179-250MG
APOSOR911179-1GEA 4050 NOK
APOSOR911179-1G APOSOR911179-250MG
(3-Propoxyphenyl)acetic acid 98%
(3-Propoxyphenyl)acetic acid
Formel: C₁₁H₁₄O₃
MW: 194.227 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD07754771
CAS-nummer: 900704-18-9

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR