(R)-(+)-2-(4-Hydroxyphenoxy)propionic acid 98%

Leverandør: Apollo Scientific

Synonymer: (R)-2-(4-Hydroxyphenoxy)propanoic acid

OR911332-500G
APOSOR911332-500GEA 1250 NOK
APOSOR911332-500G
(R)-(+)-2-(4-Hydroxyphenoxy)propionic acid 98%
(R)-(+)-2-(4-Hydroxyphenoxy)propionic acid
Formel: C₉H₁₀O₄
MW: 182.18 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00274088
CAS-nummer: 94050-90-5

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR