4-Butylcyclohexanecarboxylic acid (cis and trans mixture) 98%

Leverandør: Apollo Scientific

OR911645-1G OR911645-25G OR911645-5G
APOSOR911645-1GEA 1250 NOK
APOSOR911645-1G APOSOR911645-25G APOSOR911645-5G
4-Butylcyclohexanecarboxylic acid (cis and trans mixture) 98%
4-Butylcyclohexanecarboxylic acid (cis and trans mixture)
Formel: C₁₁H₂₀O₂
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00460757
CAS-nummer: 71101-89-8

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR