2,2-Dimethyl-3-hexanone 95%

Leverandør: Apollo Scientific

Synonymer: 2,2-Dimethylhexan-3-one

OR912109-5G
APOSOR912109-5GEA 3000 NOK
APOSOR912109-5G
2,2-Dimethyl-3-hexanone 95%
2,2-Dimethyl-3-hexanone
Formel: C₈H₁₆O
MW: 128.21 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00027140
CAS-nummer: 5405-79-8

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR