6-Methoxy-1H-indole-3-carboxylic acid 97%

Leverandør: Apollo Scientific

Synonymer: 6-Methoxy-3-indolecarboxylic Acid

OR913080-5G
APOSOR913080-5GEA 1650 NOK
APOSOR913080-5G
6-Methoxy-1H-indole-3-carboxylic acid 97%
6-Methoxy-1H-indole-3-carboxylic acid
Formel: C₁₀H₉NO₃
MW: 191.183 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD05664010
CAS-nummer: 90924-43-9

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR