4-Chloro-1H-indole-3-carboxylic acid 95%

Leverandør: Apollo Scientific

OR913259-1G
APOSOR913259-1GEA 1250 NOK
APOSOR913259-1G
4-Chloro-1H-indole-3-carboxylic acid 95%
4-Chloro-1H-indole-3-carboxylic acid
Formel: C₉H₆ClNO₂
MW: 195.61 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD07776617
CAS-nummer: 23872-36-8

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR