(R)-3-Amino-3-(4-methoxyphenyl)propionic acid 97%

Leverandør: Apollo Scientific

Synonymer: (R)-3-Amino-3-(4-methoxyphenyl)propanoic acid

OR913679-1G
APOSOR913679-1GEA 1850 NOK
APOSOR913679-1G
(R)-3-Amino-3-(4-methoxyphenyl)propionic acid 97%
(R)-3-Amino-3-(4-methoxyphenyl)propionic acid
Formel: C₁₀H₁₃NO₃
MW: 195.215 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD01318245
CAS-nummer: 131690-57-8

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR