1-Chloro-2,3-dimethoxybenzene 95%

Leverandør: Apollo Scientific

OR914653-1G
APOSOR914653-1GEA 3950 NOK
APOSOR914653-1G
1-Chloro-2,3-dimethoxybenzene 95%
1-Chloro-2,3-dimethoxybenzene
Formel: C₈H₉ClO₂
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00191399
CAS-nummer: 90282-99-8

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR