cis-3-Hexenyl acetate 97%

Leverandør: Apollo Scientific

OR916315-25G
APOSOR916315-25GEA 985 NOK
APOSOR916315-25G
cis-3-Hexenyl acetate 97%
cis-3-Hexenyl acetate
Formel: C₈H₁₄O₂
MW: 142,20 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00036563
CAS-nummer: 3681-71-8

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR