2,6-Dichloropyridine-4-methylamine 95%

Leverandør: Apollo Scientific
Warning

Synonymer: 4-(Aminomethyl)-2,6-dichloropyridine

OR9190-250MG
APOSOR9190-250MGEA 735 NOK
APOSOR9190-250MG
2,6-Dichloropyridine-4-methylamine 95%
2,6-Dichloropyridine-4-methylamine
Formel: C₆H₆Cl₂N₂
MW: 177,03 g/mol
Smeltepunkt: 73 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD06212867
CAS-nummer: 88579-63-9

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR