Etanolamin 99%

Leverandør: Apollo Scientific
Danger

OR921532-100G OR921532-1KG
APOSOR921532-100GEA 270 NOK
APOSOR921532-100G APOSOR921532-1KG
Etanolamin 99%
Etanolamin
Formel: NH₂CH₂CH₂OH
Kokepunkt: 170 °C
Smeltepunkt: 10…11 °C
Tetthet: 1,012
Flash punkt: 91 °C
MDL nummer: MFCD00008183
CAS-nummer: 141-43-5
FN: 2491
ADR: 6.1,III

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR