Propyl 4-hydroksybenzoat 95%

Leverandør: Apollo Scientific

Synonymer: Propyl 4-hydroxybenzoate, Propyl parahydroxybenzoate, Propylparaben, 4-Hydroxybenzoic acid propyl ester

OR923454-100G
APOSOR923454-100GEA 665 NOK
APOSOR923454-100G
Propyl 4-hydroksybenzoat 95%
Propyl 4-hydroksybenzoat
Formel: C₁₀H₁₂O₃
MW: 180,2 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002354
CAS-nummer: 94-13-3

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR