Dibutyl sulphate 95%

Leverandør: Apollo Scientific
Danger

OR924973-5G OR924973-25G OR924973-100G
APOSOR924973-5GEA 895 NOK
APOSOR924973-5G APOSOR924973-25G APOSOR924973-100G
Dibutyl sulphate 95%
Dibutyl sulphate
Formel: C₈H₁₈O₄S
Kokepunkt: 115…116 °C
Tetthet: 1,06
MDL nummer: MFCD00059420
CAS-nummer: 625-22-9
FN: 2810
ADR: 6.1,III

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR