Diethylamine hydrochloride 99%

Leverandør: Apollo Scientific
Warning

OR925271-5KG OR925271-1KG OR925271-250G
APOSOR925271-5KGEA 6800 NOK
APOSOR925271-5KG APOSOR925271-1KG APOSOR925271-250G
Diethylamine hydrochloride 99%
Diethylamine hydrochloride
Formel: C₄H₁₂ClN
Kokepunkt: 320…330 °C/760mmHg
Smeltepunkt: 227…230 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00012499
CAS-nummer: 660-68-4

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR