Indane-1-acetic acid 95%

Leverandør: Apollo Scientific

Synonymer: 2-(2,3-Dihydro-1H-inden-1-yl)acetic acid

OR925275-1G
APOSOR925275-1GEA 2300 NOK
APOSOR925275-1G
Indane-1-acetic acid 95%
Indane-1-acetic acid
Formel: C₁₁H₁₂O₂
MW: 176,215 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00156939
CAS-nummer: 38425-65-9

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR