3-(4-Carboxyphenyl)propionic acid 98%

Leverandør: Apollo Scientific

Synonymer: 4-(2-Carboxyethyl)benzoic Acid

OR925442-25G
APOSOR925442-25GEA 2075 NOK
APOSOR925442-25G
3-(4-Carboxyphenyl)propionic acid 98%
3-(4-Carboxyphenyl)propionic acid
Formel: C₁₀H₁₀O₄
MW: 194.19 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00016560
CAS-nummer: 38628-51-2

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR