(4-Propoxyphenyl)acetic acid 98%

Leverandør: Apollo Scientific

Synonymer: 2-(4-Propoxyphenyl)acetic acid

OR925627-1G OR925627-5G
APOSOR925627-1GEA 1875 NOK
APOSOR925627-1G APOSOR925627-5G
(4-Propoxyphenyl)acetic acid 98%
(4-Propoxyphenyl)acetic acid
Formel: C₁₁H₁₄O₃
MW: 194.23 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00460580
CAS-nummer: 26118-57-0

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR