3,4-Dihydro-2H-quinoline-1-carboxylic acid methyl ester 95%

Leverandør: Apollo Scientific

OR926745-1G OR926745-5G
APOSOR926745-1GEA 2350 NOK
APOSOR926745-1G APOSOR926745-5G
3,4-Dihydro-2H-quinoline-1-carboxylic acid methyl ester 95%
3,4-Dihydro-2H-quinoline-1-carboxylic acid methyl ester
Formel: C₁₁H₁₃NO₂
MW: 191.226 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD03364950
CAS-nummer: 94567-78-9

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR