Ac-HMBA-linker 98%

Leverandør: Apollo Scientific

Synonymer: 4-(Acetoxymethyl)benzoic acid, 4-Acetoxymethylbenzoic acid

OR928683-25G
APOSOR928683-25GEA 5650 NOK
APOSOR928683-25G
Ac-HMBA-linker 98%
Ac-HMBA-linker
Formel: C₁₀H₁₀O₄
MW: 194.2 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD01090957
CAS-nummer: 15561-46-3

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR