1,1-Diphenylethanol 95%

Leverandør: Apollo Scientific

Synonymer: α-Methylbenzhydryl alcohol

OR930645-100G
APOSOR930645-100GEA 1700 NOK
APOSOR930645-100G
1,1-Diphenylethanol 95%
1,1-Diphenylethanol
Formel: (C₆H₅)₂C(OH)CH₃
MW: 198.26 g/mol
Kokepunkt: 328
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004450
CAS-nummer: 599-67-7

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR