4-Isobutylcyclohexanecarboxylic acid (cis and trans mixture) 95%

Leverandør: Apollo Scientific

Synonymer: 4-Isobutylcyclohexanecarboxylic acid

OR930868-5G
APOSOR930868-5GEA 4925 NOK
APOSOR930868-5G
4-Isobutylcyclohexanecarboxylic acid (cis and trans mixture) 95%
4-Isobutylcyclohexanecarboxylic acid (cis and trans mixture)
Formel: C₁₁H₂₀O₂
Lagringstemperatur: Romtemperatur
CAS-nummer: 38792-88-0

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR