1-Methyl-1H-indazole-7-carbaldehyde 98%

Leverandør: Apollo Scientific
Warning

OR931136-1G
APOSOR931136-1GEA 1625 NOK
APOSOR931136-1G
1-Methyl-1H-indazole-7-carbaldehyde 98%
1-Methyl-1H-indazole-7-carbaldehyde
Formel: C₉H₈N₂O
MW: 160,173 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD11111019
CAS-nummer: 951030-58-3

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR