Calcium lactate gluconate 97%

Leverandør: Apollo Scientific

Synonymer: rac-(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate, 2-hydroxypropanoate, Calcium

OR931348-100G
APOSOR931348-100GEA 1250 NOK
APOSOR931348-100G
Calcium lactate gluconate 97%
Calcium lactate gluconate
Formel: Ca·C₆H₁₁O₇·C₃H₅O₃
Smeltepunkt: 140 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD01320836
CAS-nummer: 11116-97-5

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR