Isobutyl isovalerate 97%

Leverandør: Apollo Scientific

Synonymer: Isobutyl Isovalerate, Isovaleric acid Isobutyl ester

OR932977-500G
APOSOR932977-500GEA 2775 NOK
APOSOR932977-500G
Isobutyl isovalerate 97%
Isobutyl isovalerate
Formel: C₉H₁₈O₂
MW: 158.2380 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00048349
CAS-nummer: 589-59-3

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR