cis-4-tert-Butylcyclohexanecarboxylic acid 95%

Leverandør: Apollo Scientific

OR936255-10G OR936255-1G OR936255-5G
APOSOR936255-10GEA 10900 NOK
APOSOR936255-10G APOSOR936255-1G APOSOR936255-5G
cis-4-tert-Butylcyclohexanecarboxylic acid 95%
cis-4-tert-Butylcyclohexanecarboxylic acid
Formel: C₁₁H₂₀O₂
Lagringstemperatur: Romtemperatur
CAS-nummer: 943-28-2

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR