(S)-(-)-β-Citronellol 95%

Leverandør: Apollo Scientific
Warning

OR936732-25G OR936732-5G OR936732-100G
APOSOR936732-25GEA 965 NOK
APOSOR936732-25G APOSOR936732-5G APOSOR936732-100G
(S)-(-)-β-Citronellol 95%
(S)-(-)-β-Citronellol
Formel: C₁₀H₂₀O
Kokepunkt: 225…226 °C
Tetthet: 0,856 g/cm³ (25 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00063214
CAS-nummer: 7540-51-4

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR