1-(2-Cyanoethyl)-2-phenylimidazole 95%

Leverandør: Apollo Scientific

Synonymer: 3-(2-Phenyl-1-imidazolyl)propionitrile

OR937224-25G
APOSOR937224-25GEA 1900 NOK
APOSOR937224-25G
1-(2-Cyanoethyl)-2-phenylimidazole 95%
1-(2-Cyanoethyl)-2-phenylimidazole
Formel: C₁₂H₁₁N₃
MW: 197,24 g/mol
Lagringstemperatur: Romtemperatur
CAS-nummer: 23996-12-5

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR