2-Chloro-6-methylbenzonitrile 95%

Leverandør: Apollo Scientific

OR937301-5G
APOSOR937301-5GEA 2000 NOK
APOSOR937301-5G
2-Chloro-6-methylbenzonitrile 95%
2-Chloro-6-methylbenzonitrile
Formel: C₈H₆ClN
MW: 151,60 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00001782
CAS-nummer: 6575-09-3

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR