3,5-Dimetylsykloheksanon 95%

Leverandør: Apollo Scientific

OR938239-5G
APOSOR938239-5GEA 915 NOK
APOSOR938239-5G
3,5-Dimetylsykloheksanon 95%
3,5-Dimetylsykloheksanon
Formel: C₈H₁₄O
MW: 126.1962 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00019467
CAS-nummer: 2320-30-1

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR